Print…
GrowDex®-A hydrogel
Fornecedor: UPM Biomedicals
300.103.002 300.103.005 300.103.010 300.103.305
300.103.002EA 170 EUR
300.103.002 300.103.005 300.103.010 300.103.305
GrowDex®-A hydrogel
Cultura celular, Suplementos e aditivos
GrowDex®-A is a ready to use, transparent hydrogel that has been specifically developed to bind different biotinylated molecules whose presence may enhance the cell culture conditions for specific 3D cell culture applications.
  • 1.0% solution (w/v) of avidin conjugated nanofibrillar cellulose, 99% purified water
  • Transparent, not autofluorescent, suitable for microscopy studies
  • Devoid of endotoxins and pathogens
  • Biocompatible with human cells and tissues
  • Sterile, tested and found negative for bacteria, fungi, and mycoplasma
  • Cell viability confirmed/tested using HepG2 cells
  • Viscosity confirmed by rheology tests
GrowDex®-A is a transparent hydrogel that consists of avidin conjugated nanofibrillar cellulose (1.0% w/v) and 99.0% purified water that is supplied sterile. The binding capacity of biotinylated molecules e.g. proteins, peptides, antibiotics makes GrowDex-A ideal for use different cell culture applications, such as spheroid formation. GrowDex-A is ready to use: first mix with the desired biotinylated molecule and then add cells and culture media, no gelation or cross-linking step required. Ambient handling and shear thinning properties of GrowDex-A enable easy dispensing and use in automated systems.
Please refer to the product Instructions for Use and the Quickstart Guide and videos for guidance on use of the product and assay optimization.
Embalagem: Supplied in syringe packaging for simple, accurate dispensing.
Atenção: GrowDex®-A is supplied at a concentration of 1.0%, it is not a concentrate. For Research Use Only. Not for use in diagnostic or therapeutic procedures.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Espécies
Origem