Print…

Anti-IgM (µ chain) Rabbit Polyclonal Antibody (Biotin)

Fornecedor: Tonbo Biosciences
30-8075-M001
30-8075-M001EA 374 EUR
30-8075-M001
Anti-IgM (µ chain) Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Rabbit anti-Human IgM (mu chain) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR