Print…
CF™ dye streptavidin and monoclonal mouse anti-biotin conjugates
Fornecedor: Biotium
29031 29032 29033 29034 29035 29036 29037 29038 29039 29040 29030
29031.EA 323 EUR
29031. 29032. 29033. 29034. 29035. 29036. 29037. 29038. 29039. 29040. 29030.
CF™ dye streptavidin and monoclonal mouse anti-biotin conjugates
Sistemas de ligação e rotulagem de proteínas
CF™ dyes are superior to other fluorescent dyes for protein labelling due to advantages in brightness, photostability and water solubility.
Streptavidin conjugates are typically used as secondary reagents to detect biotinylated probes such as primary antibodies for flow cytometry, Western blotting, immunofluorescence staining and other applications.
Streptavidin conjugates are supplied in lyophilised solid format. After reconstitution with 0,5 ml PBS, streptavidin concentration is 2 mg/ml with 0,01% sodium azide. Monoclonal mouse anti-biotin antibody conjugates are supplied at 2 mg/ml in PBS pH 7,4 containing 50% glycerol, 2 mg/ml BSA (IgG-free and protease-free), and 0,05% sodium azide.