Print…
Ion exchange chromatography column, HiScreen™ Capto™ adhere
Fornecedor: Cytiva
28-9269-81
28-9269-81EA 248 EUR
28-9269-81
Ion exchange chromatography column, HiScreen™ Capto™ adhere
Cromatografia, Colunas
HiScreen Capto adhere columns are prepacked with Capto adhere ion exchange chromatography resins, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.
  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer
Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Pressão máx. 3 bar
pH range 3 - 12