Print…
MEM vitamin solution, 100X, Corning®
Fornecedor: Corning
25-020-CI
25-020-CIEA 111.6 EUR
25-020-CI
MEM vitamin solution, 100X, Corning®
Cultura celular, Suplementos e aditivos
The addition of MEM vitamins to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells.
This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.