Print…
Burette clamps, double
Fornecedor: BOCHEM
5600
241-3169EA 84.8 EUR
241-3169
Burette clamps, double
Pinças /Clamps Pinças de uso geral
Aluminium.
  • With bosshead for fixing to retort stand with Ø 0 to 13
  • With PVC rolls
  • Distance between rolls 170 mm