Print…
Scissors, pointed tips, Rubis®
Fornecedor: OUTILS RUBIS
1C300/EL
233-0219EA 31.9 EUR
233-0219
Scissors, pointed tips, Rubis®
Tesouras
High precision micro scissors with sharp pointed tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.