Print…
Double spatulas, powder
Fornecedor: RSG
037.185 037.130 037.150 037.210
231-1411EA 5.5 EUR
231-1411 231-0544 231-0545 231-0546
Double spatulas, powder
Espátulas
Stainless steel.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Very good thermal resistance, autoclavable
  • Good chemical resistance