Print…
Tubing clamps, Kocher (Ochsner) type
Fornecedor: HAMMACHER KARL
HSE094-20
231-0251EA 117 EUR
231-0251
Tubing clamps, Kocher (Ochsner) type
Pinças /Clamps Pinças para tubos
Stainless steel, straight. Ideal for stopping or controlling liquid flow rates in tubing connections.
  • Very good thermal resistance, autoclavable
  • Corrosion-resistant
  • Antimagnetic