Print…
Apiezon® sealing compound Q
Fornecedor: M I MATERIALS
FAP/W/Q-01000
22844.298EA 405 EUR
22844.298
Apiezon® sealing compound Q
Lubrificantes e gorduras
Used to seal joints and gaps and to effect temporary repairs. Good for sealing flat unground joints. Can be moulded by hand as it is a putty-like substance. For use at ambient temperatures.
  • Vapour pressure 1 x 10-4 torr at 20°C