Print…
Sample vials in Lab File®, WHEATON®
Fornecedor: DWK Life Sciences
224801 224802 224804 224814 224831 224833 224805 224806 224808 W224815
218-2221EA 133 EUR
218-2221 218-2222 218-2223 218-2230 218-2201 218-2203 218-2224 218-2225 218-2226 WHEAW224815
Sample vials in Lab File®, WHEATON®
Vials
For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.
Informações de encomenda: Without cap.