Print…
Stoppers, Glass, Conical Ground Joint
Fornecedor: DWK Life Sciences
216220708
217-0767EA 211 EUR
217-0767
Stoppers, Glass, Conical Ground Joint
Rolhas Rolhas de vidro
DURAN® glass stopper with conical ground joint, for reagent bottles and oxygen bottles according to Winkler.
  • Made of borosilicate 3.3 glass