Print…
Wire baskets
Fornecedor: BOCHEM
10050 10051 10052 10053 10054 10055 10057 10056
216-2840EA 77.7 EUR
216-2840 216-2841 216-2842 216-2843 216-2844 216-2845 216-2846 BOCH10056
Wire baskets
Cestos Cestos metálicos
Stainless steel.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.