Print…
Nalgene® Certified Platinum Clean HDPE Bottles and Carboys, Sterile
Fornecedor: Thermo Fisher Scientific
CE-N2289-0075 CE-N2289-0020 CE-N2289-0050 CE-N2099-0032 CE-N2199-0016 CE-N2199-0032 CE-N2099-0016 CE-E2099-0001 CE-N2099-0008 CE-N2199-0008
216-2309EA 1290 EUR
216-2309 216-2307 216-2308 216-2303 216-2305 216-2306 216-2300 216-2298 216-2299 216-2304
Nalgene® Certified Platinum Clean HDPE Bottles and Carboys, Sterile
Frascos Garrafões
These certified platinum clean HDPE bottles and carboys help reduce the potential for carry-over contamination in biopharmaceutical and vaccine productions.
  • Sterile and ready to use
  • Washed with USP water
  • Lot-to-lot tested
  • Robust and versatile
These products include particulate certifications to under 1/3 the allowable limits according to USP <788> and lower endotoxin levels of 0,25 EU/ml, to help mitigate potential sources of unwanted impurities.
All container/closure systems certified leakproof to protect precious contents.