Print…
VWR®, Garrafões de boca larga, redondos, com tampa de rosca
Fornecedor: VWR Collection
216-1708EA 38.6 EUR
216-1708 216-1730 216-1733 216-1919
VWR®, Garrafões de boca larga, redondos, com tampa de rosca
Frascos Garrafões
LDPE ou PP, translúcidos com tampas de rosca roxas.
Adequado para armazenar e dispensar soluções e meios. Dispõe de uma pega integrada. Graduado para identificação do volume dispensado.
LDPE carboys
Ideal for high-purity applications, as they have high resistance to chemicals (widely used in ophthalmic products like eye droppers and nasal sprays).
Preferred resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impacts (will not break even if it falls down accidentally, whereas PP carboys may lead to cracking at negative temperatures)
LDPEMais informação do produto