Print…
VWR®, Garrafões, de boca larga, PP
Fornecedor: VWR Collection
216-0616EA 122 EUR
216-0616 216-0617 216-0618 216-0623 216-0624 216-0625 216-0630 216-0631 216-0636 216-0637 216-0638
VWR®, Garrafões, de boca larga, PP
Frascos Garrafões
PP, opaco.
  • A tecnologia Versatile Cap assegura a flexibilidade dos conectores intercambiáveis
  • Versão de boca extralarga para facilitar o enchimento e a limpeza
  • A tecnologia Smart Barb oferece opções de torneiras intercambiáveis
The carboy is an advanced fluid handling system developed with the end user in mind. This ergonomically-designed carboy body, coupled with VWR® Versatile Cap technology, provides the user with something that is easy to use and easy to clean. Made from polypropylene that offers good chemical resistance and heat stability.Mais informação do produto