Print…
VWR®, Frascos, de boca larga, com tampa roscada
Fornecedor: VWR Collection
215-5685EA 77.7 EUR
215-5685 215-5684 215-5683 215-5682 215-5681
VWR®, Frascos, de boca larga, com tampa roscada
Frascos Frascos de Plástico
PP, translúcido.
  • Very good chemical resistance
  • Heat resistant up to +121 °C (for 20 minutes), prolonged use up to +100 °C
  • Leakproof
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The wide opening allows easy filling. Can be filled with hot substances. Come with easy to read blue graduations.