Print…
VWR® Aspirator Bottles, with Stopcock
Fornecedor: VWR Collection
215-4179EA 126 EUR
215-4179 215-4180 215-4181 215-4182
VWR® Aspirator Bottles, with Stopcock
Frascos Frascos com Torneira
Borosilicate 3.3 glass, with blue PP screw cap (GL 45) and interchangeable glass stopcock.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • All parts of the bottle are fully autoclavable
Ideal for the storage of liquids containing aggressive acids or alkalis.