Print…
Wash bottles, integral, wide neck, multilingual, Azlon®
Fornecedor: SciLabware
WGW801P WGW802P WGW803P WGW804P WGW805P
215-4163EA 81.1 EUR
215-4163 215-4164 215-4165 215-4166 215-4167
Wash bottles, integral, wide neck, multilingual, Azlon®
Frascos Esguichos
Multilingual, wide neck wash bottles with coloured caps.
  • Indelibly printed in English, French, German and Spanish
  • Fitted with colour coded one-piece PP closure that maintains it's shape in use
  • Vented cap with PTFE membrane allows venting and prevents pressure build up
  • Designed with integral spout to prevent contamination
  • Good chemical resistance
This round wash bottle is moulded with an integral spout so you can say goodbye to contamination caused by the inner tube touching the bench top during re-filling. Choose from five solvent labels all includingMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable