Print…
Jacketed beakers, tall form
Fornecedor: SciLabware
1025/22SC 1025/18SC 1025/28SC 1025/10SC
213-0042EA 483 EUR
213-0042 213-0052 213-0054 213-0373
Jacketed beakers, tall form
Copos
PYREX®, borosilicate glass, without spout.
  • Very good chemical resistance
  • Very good thermal resistance
  • With screw thread and plastic threaded tubing connectors
  • Tube nozzles with 11 mm ext. diameter (Ø) for 250 and 600 ml beakers and 14 mm ext. diameter (Ø) for 1000 and 2000 ml beakers
For maintaining liquid samples at a constant temperature. The temperature controlling medium is pumped through the double-walled jacket by means of a recirculating thermostat.