Print…
Test tubes, with rim, heavy wall
Fornecedor: MURRAY S
G032/01 G032/02 G032/03 G032/05 G032/06 G032/08 G032/10
212-7035EA 80.7 EUR
212-7035 MURRG032/02 MURRG032/03 212-7036 212-7037 MURRG032/08 212-7038
Test tubes, with rim, heavy wall
Tubos Tubos para ensaio
Borosilicate test tubes are suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples. They are also highly chemically resistant.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes