Print…
VWR® Mega, Suportes para tubos de ensaio
Fornecedor: VWR Collection
211-0366EA 70.1 EUR
211-0366 211-0363 211-0364 211-0365 212-0660 212-0661 212-0662 212-0663
VWR® Mega, Suportes para tubos de ensaio
Suportes/Racks Tube Racks
High capacity storage for your tubes.
  • Sturdy, material for easy and secure transport of blood collection and other tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
  • Autoclavable
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable