Print…
Vacuum flask, Quickfit®
Fornecedor: SciLabware
FEH250/3
212-0588EA 40.6 EUR
212-0588
Vacuum flask, Quickfit®
Balões Frascos de Cultura
Made of Borosilicate glass, suitable for use under vacuum. With ground glass socket.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With heavy walls and bases. Suitable for use under vacuum.