Print…
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Fornecedor: Merck Millipore Calbiochem
201102-100
201102-100EA 265 EUR
201102-100
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Sistemas de ligação e rotulagem de proteínas
Bilirubin conjugate, ditaurate is suitable as a direct bilirubin standard. It behaves similarly to purified bilirubin glucuronides from human bile in diazo tests.
Note: Unstable in solution.