Print…
Claisen type distillation heads
Fornecedor: Lenz Laborglas GmbH & CO.KG
05030033 05030044
201-1174EA 53.5 EUR
201-1174 201-2226
Claisen type distillation heads
Componentes para destilação Cabeças de Destilação
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Acc. to DIN 12594 and in addition to standard, with two parallel NS 14/23 sockets and two NS cones (descending angle: 75°), for distillation thermometers of 55 mm built-in length.