Print…
Lab tower, LT 828
Fornecedor: KNF
LT828
181-3110EA 0 EUR
181-3110
Lab tower, LT 828
Bombas Bombas de Diafragma
Multipurpose laboratory instrument for handling liquids. It can be used for filtering (in combination with a vacuum filtration unit), pipetting and aspirating liquids. The flow rate during aspirating and filtering, and the aspiration and dispensing speed when pipetting, can both be regulated from the control panel. A vacuum gauge indicates the pressure in the collecting bottle. The vacuum that the tower needs in order to work is created by a diaphragm vacuum pump. Liquid media are prevented from penetrating the pump by a protective sterile filter. The tower is equipped with locking wheelsMais informação do produto