Print…
Fluid aspiration systems, biovac 106
Fornecedor: Welch by Gardner Denver
112037 112037-02 112037-04
181-0176EA 1950 EUR
181-0176 181-0587 181-0588
Fluid aspiration systems, biovac 106
Bombas Bombas de Tambor
With the biovac 106, nonflammable fluids can be removed easily and very precisely from slides, Petri dishes, cell culture containers, etc, by using different pipettes or glass tips and Pasteur pipettes which can be easily connected to the handvac pipette. The biovac 106 is switched on and off directly at the device or via the optionally available foot switch.
  • Resistant diaphragm pump
  • Operation via ON/OFF switch or optional foot switch
  • Autoclavable receiver is available in 2 L glass or 4 L polypropylene
  • Power supply: 115/230 V, 50/60 Hz