Print…
Cation exchange chromatography columns, HiTrap™ SP FF
Fornecedor: Cytiva
17-5054-01 17-5157-01
17-5054-01EA 172 EUR
17-5054-01 17-5157-01
Cation exchange chromatography columns, HiTrap™ SP FF
Cromatografia, Colunas
HiTrap™ SP FF is pre-packed with SP Sepharose™ Fast Flow, and is a strong cation exchanger for small scale protein purifications as well as screening of binding and elutions conditions.
  • Fast, easy ion exchange separations either alone or connected in series
  • High flow rates and good scale-up potential
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
  • Predictable scale-up
Q, SP, DEAE, and CM Sepharose™ Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Pressão máx. 5 bar
pH range 4 - 13