Print…
Affinity chromatography columns, HiTrap™ Heparin HP
Fornecedor: Cytiva
17-0406-01 17-0407-01 17-0407-03
17-0406-01EA 298 EUR
17-0406-01 17-0407-01 17-0407-03
Affinity chromatography columns, HiTrap™ Heparin HP
Cromatografia, Colunas
HiTrap Heparin HP columns are designed for high-resolution purification of DNA-binding proteins, coagulation factors, and other plasma proteins.
  • Suitable for purification of antithrombin III, coagulation factors, and other plasma proteins
  • Useful for screening and optimisation before scaling up to bulk resins such as Heparin 6 Fast Flow
  • High-resolution purification and elution in narrow peaks, minimizing the need for further concentration steps
  • Prepacked with Heparin Sepharose High Performance resin
  • HiTrap format compatible with syringes, pumps, and
Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Pressão máx. 5 bar
pH range 5 - 10
Armazenamento 4 to 30 °C, 20% Ethanol