Print…
Tubos para medição de gases, para bomba Accuro
Fornecedor: Dräger
6718501 6718701 6722101 6723301 6728521 6728531 6728651 6733181 8101641 CH20001 6730201 8101751 CH25001 6728081 6728311 6728371 8101341 8101491 CH23101 8101511 6733141 8103261 8101121 6722701 6733031 8101061 8101691 CH20201 CH21001 8101721 6728861 6728451 6728161 8101141 6730501 8101351 CH31801 6718401 6728111 6733171 CH31201 6728761 6728051 8101681 6728681 CH24101 6724501 6728381 8103351 CH19601 CH25803 6728461 CH20401 8101021 8103591 CH19801 CH21101 CH25903 CH26303 8103601 8103631 6728511 8103421 6724701 8103671 8103701 6718601
151-0218EA 187 EUR
151-0218 151-0137 151-0141 151-0143 151-0156 151-3157 151-0158 151-0165 151-3149 151-0219 151-3162 151-0178 151-0196 151-0149 151-0103 151-0104 151-0110 151-0172 151-3158 151-3182 151-0163 111-0234 151-0167 151-0142 151-0161 118-0305 151-0174 118-0334 118-0336 151-0175 151-0107 118-0295 118-0291 151-0168 118-0302 118-0307 118-0351 118-0284 118-0290 118-0303 118-0350 118-0299 118-0288 118-0314 118-0298 118-0340 118-0285 118-0293 118-0323 118-0332 118-0341 118-0296 118-0335 118-0304 118-0356 118-0333 118-0337 118-0342 118-0343 118-0392 118-0331 151-0155 118-0354 118-0352 DRAE8103671 DRAE8103701 DRAE6718601
Tubos para medição de gases, para bomba Accuro
Tubos de gás
Different gases and vapours can be detected and measured such as concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.