Print…
Gas detection tubes for ethylene oxide
Fornecedor: Dräger
6728961 6728241
151-0108EA 111 EUR
151-0108 118-0292
Gas detection tubes for ethylene oxide
Tubos de gás
To measure the 'spot concentrations' of ethylene oxide.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.