Print…
Glass cleaners, SURE®
Fornecedor: Diversey
100891496
141-1570EA 33.6 EUR
141-1570
Glass cleaners, SURE®
Soluções de limpeza Detergentes e Limpadores de Superfícies
Ready to use spray cleaner for streak-free glass cleaning.
  • Excellent removal of fingermarks as well as the spot treatment of other slightly adherent soils
  • Easy spray-on, wipe-off action
  • No fragrance nor dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product for user
SURE® glass cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to theMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable Materiais sustentáveis,Maior segurança para a saúde e ambiente,Embalagem sustentável