Print…
asecos, armários de armazenamento de segurança, S-Classic
Fornecedor: asecos
30115001 30115002 30116001 30116002 30238001 30238002 30239001 30239002 30240001 30121001 30121002 30122001 30122002 30204001 30204002 30205001 30205002 30227001 30227002 30228001 30228002 30229001 30229002 30230001 30230002 30240002 30241001 30241002 32533005 32532005
135-1759EA 3177 EUR
135-1759 135-1760 135-2564 135-2565 135-2576 135-2577 135-2578 135-2579 135-2580 135-2067 135-2068 135-2566 135-2567 135-1761 135-1762 135-2568 135-2569 135-1697 135-1695 135-2570 135-2571 135-2572 135-2573 135-2574 135-2575 135-2581 135-2582 135-2583 135-2586 135-2585
asecos, armários de armazenamento de segurança, S-Classic
Mobiliário Armários/Armários Segurança
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
  • Robust construction and longevity - safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
  • Convenient handling: Doors can be opened with minimal effort
  • User friendly and safe: Door open arrest system (optional), convenient and safe removal or storage of containers
  • No unauthorised use: Doors lockable with cylinder locking (integration in a locking system possible)
  • Easy alignment: Adjusting aids to compensate for
Mais informação do produto