Print…
Acessórios de ventilação, sistemas de extracção
Fornecedor: DUPERTHAL
2.00.175EX 2.00.360 2.00.079 2.00.080 2.00.081 2.00.339-1 2.00.320-1 2.00.397-1S
132-0621EA 645 EUR
132-0621 132-0626 132-0735 DUEH2.00.080 DUEH2.00.081 DUEH2.00.339-1 DUEH2.00.320-1 DUEH2.00.397-1S
Acessórios de ventilação, sistemas de extracção
Ventiladores
Exhaust air monitor optional with ventilator: Easy ventilation and monitoring of the venting system for safety cabinets. A sensor monitors the air flow speed/stream and gives a visual or audible signal if the value drops below the set-point value. The complete exhaust monitoring unit is supplied pre-assembled (ready to plug in) and is placed directly on the cabinet. The set-point value for the air stream flow rate is preset in the factory and can be individually adjusted by the pressure difference socket. In addition to the volumetric flow rate the power supply is also monitored. The systemMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable