Print…
BRUTE® Vented Wheelie Bin, 167 L Container without Lid
Fornecedor: Rubbermaid Commercial Products
2131929
129-3981EA 318 EUR
129-3981
BRUTE® Vented Wheelie Bin, 167 L Container without Lid
Caixas e contentores para resíduos Baldes do lixo
Famed durability, with wheels. Reduce injury risk and drive waste efficiency in the toughest environments.
  • Ergonomic handle is designed to require less force when moving loads up to 113 kg, with DuraTread never-flat wheels
  • Handle design enables users to maintain proper back posture by leveraging shoulder and torso muscles rather than relying on arm muscles alone
  • Integrated tilt and kickplate provides maximum tilting when transporting materials
  • Venting channels allow for air flow around and below the bag, making for low force liner removal that is reduced
Mais informação do produto