Print…
Polypropylene Bowl Covers
Fornecedor: KEYSTONE CLEANROOM PRODUCTS
BWLPPW24E BWLPPW26E BWLPPW30E BWLPPW36E BWLPPW42E BWLPPW48E BWLPPW72E
129-2871EA 1290 EUR
129-2871 129-2872 129-2873 129-2874 129-2875 129-2876 129-2877
Polypropylene Bowl Covers
Capas de proteção
Keeps cleaned, non sterile production equipment surfaces protected from external sources of contamination; or use ‘in process’ to prevent cross contamination of products.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear- and puncture-resistant non woven polypropylene
  • Individually poly bagged
These polypropylene bowl covers are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover or used 'in process' to prevent product cross contamination. Individually polyMais informação do produto