Print…
Rubbermaid® Untouchable® Containers
Fornecedor: Rubbermaid Commercial Products
FG395873BLUE FG356973BLUE FG356988BLA FG352000GRAY FG395800GRAY FG354600GRAY FG356988GRAY
129-1834EA 209 EUR
129-1834 129-1838 129-1837 129-1790 129-1835 129-1841 129-2469
Rubbermaid® Untouchable® Containers
Caixas e contentores para resíduos Baldes do lixo
Stationary containers provide convenient central collection sites for multiple work stations or areas.
  • Venting channels make removing liners up to 50% easier, improving productivity
  • Durable and crack-resistant, even under tough conditions
  • Provided with a 5 year warranty
Bag cinch secures liner around the rim of the container and allows for quick, knot-free liner changes.
Acessórios: Untouchable® tops (sold separately) help facilitate recycling sortation and waste disposal.