Print…
Wipes, pre-saturated with silicone, SilSat™
Fornecedor: MICRONOVA
SSW6-01IR
129-0248EA 931 EUR
129-0248
Wipes, pre-saturated with silicone, SilSat™
Toalhetes Toalhetes e toalhas descartáveis Toalhetes pré-saturados
SilSat wipes address the problem of using lubricant in a controlled application by offering high grade silicone on a pre-saturated wipe. Each polyester wipe carries several grams of silicone, enough to service gears or rollers on fill lines or lubricate stopper caps and valves. Wipes are gamma irradiated.
  • High grade silicone/alcohol blend
  • Light impregnation
  • Avoids possible transfer of silicon into product or other critical surfaces
Cor:branco
Weight: 106 g/m²
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable