Print…
PIG® HazMat mat pads and rolls
Fornecedor: NEW PIG
MAT301 MAT302 MAT309 MAT310 MAT351 MAT354 MAT3510 MAT303
121-0527EA 148.75 EUR
121-0527 121-0031 121-0528 121-0529 121-0531 NEWPMAT354 121-0099 121-0156
PIG® HazMat mat pads and rolls
Materiais para controlo de derrames
Specially treated mats that are ideal for absorbing highly concentrated corrosives like 98% sulphuric acid and 30% sodium hydroxide.
  • Absorbs the widest range of acids, caustics and unknown liquids - even those with high concentrations like 98% sulphuric acid and 30% sodium hydroxide
  • Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Eight layers of 100% PP are thermally bonded to make PIG mats exceptionally strong; they won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of
Mais informação do produto