Print…
PIG® Essentials Pro chemical mat pads
Fornecedor: NEW PIG
CPFH100
121-0192EA 103.5 EUR
121-0192
PIG® Essentials Pro chemical mat pads
Materiais para controlo de derrames
Chemical Mat that quickly absorbs acids, caustics and unknown liquids for effective clean-up of high volume spills.
  • 100% polypropylene mat quickly absorbs nearly any type of caustic, hazardous or unknown liquid- including acids, bases, solvents and chemicals
  • Colour-coded safety yellow for fast identification; know exactly what you're reaching for in an emergency spill situation
  • Top and bottom layers of fine fibre are built of densely woven, small diameter strands of fibre to increase surface area-enabling the mat to quickly soak up liquids on contact
Mais informação do produto