Print…
Wipes for polishing applications, VeraClean Polish Plus
Fornecedor: PGI NONWOVEN
74101 74104
115-2500EA 29 EUR
115-2500 115-2501
Wipes for polishing applications, VeraClean Polish Plus
Toalhetes Panos de limpeza reutilizáveis Panos para aplicações gerais
VeraClean Polish Plus is a bulky, velvet-soft polishing cloth that is designed for use in critical environments and on sensitive surfaces. It is suitable for use with almost all industrial polishing pastes. Due to its dense structure, VeraClean Polish Plus provides an alternative to high viscosity polish material.
  • High absorbency allows easy removal of excess polishing paste, water, oil and ink
  • Extremely soft, making it ideal for polishing
  • Resistant to most solvents and chemicals
  • Solvent-, silicone- and chemical-free
  • Low linting
Ideal for maintenanceMais informação do produto