Print…
Wipes for hand and surface cleaning, pre-saturated, Tork
Fornecedor: Essity
190592
115-0583EA 147 EUR
115-0583
Wipes for hand and surface cleaning, pre-saturated, Tork
Toalhetes Toalhetes e toalhas descartáveis Toalhetes pré-saturados
Pre-moistened wipes made from a viscose and polyester mix. The single ply wipes are tough on grease and dirt but gentle enough on skin to use several times a day without risking dryness or irritation to skin.
  • Convenient, portable and recyclable dispensing container: Wipes are easy to dispense
  • Waterless cleaning: Tough on grease and dirt
  • Kind to skin and fragrance-free
Colour: White
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable