Print…
Wipes, optical grade, Amplitude™ Optic Wipe
Fornecedor: CONTEC
C6-99
115-0579EA 32.4 EUR
115-0579
Wipes, optical grade, Amplitude™ Optic Wipe
Toalhetes Toalhetes para salas limpas Seco
These absorbent non abrasive wipes consist of long staple cellulosic fibres which are wet laid in a paper making process to form an extremely clean sheet.
  • Non woven, long staple fibre construction
  • Very low extractables level, moderate particle generation
  • Can be used as separating layers between CDs and silicon wafers and for packaging and storage of optical products
Ideal for delicate cleaning of optical products, drying critical products and for use as a lining for cleanroom surfaces or trays.
Colour: White.
Atenção: Data accurate at time ofMais informação do produto
Encomendar
ESPECIFICAÇÕES
Modelo Amplitude™
Environmentally Preferable
Peso base 16,6 g/m²
Absorção - extrínseca 79,2 ml/m²
Absorção - intrínseca 4,79 ml/g
Extraíveis - usando água desionizada 0,0238 g/m²
Extraíveis - usando álcool isopropílico 0,0110 g/m²
Particle generation (>=0,5 µm) 42,42 × 10⁶/m²
Iões - cloreto 0
Iões - sódio 199,83 ppm