Print…
Cleanroom hoods, CleanMax®
Fornecedor: LAKELAND CLEANMAX APPAREL
CTL713CM
114-9543EA 539 EUR
114-9543
Cleanroom hoods, CleanMax®
Roupas Descartáveis
Clean manufactured disposable apparel are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All apparel provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, andMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Classe da sala limpa 100/ISO 5
Gênero
Aplicações de roupa Cleanroom