Print…
Shoe covers, PP, Hopen® Safefeet® SkidGuard™, with PE sole
Fornecedor: Medicom
10.007
113-9989EA 170 EUR
113-9989
Shoe covers, PP, Hopen® Safefeet® SkidGuard™, with PE sole
Roupas Descartáveis
PP shoe covers feature a 60 μm PE plastic-coated, non-slip sole to prevent slipping and ensure a long product life.
  • Class 1 medical device
  • Single use, non-sterile
  • PP non-woven upper with PE sole
  • Made in France
These lightweight garments are adaptable in size to fit over all types of shoes. The PE sole helps to reduce the risk of abrasion on rough floors.
Ideal for use in medical and clinical areas, for food processing, and in laboratories.
Sole thickness: 60 μm
Weight: 35 g/m²