Print…
Cold storage overall, Matterhorn 6402
Fornecedor: SIOEN
6402/276/S 6402/276/M 6402/276/L 6402/276/2XL 6402/276/3XL 6402/276/1XL
113-7162EA 284 EUR
113-7162 113-7161 113-7160 113-7158 113-7159 113-7157
Cold storage overall, Matterhorn 6402
Uniformes
This overall, made of polyester, cotton and Isosoft lining, offers protection from temperatures down to −40 °C. Cold bridges are avoided.
  • Fixed quilted lining, knitted wind cuffs in sleeves
  • One inside pocket, one patched chest pocket, two patched pockets, pen pocket
  • Fleece inside collar, zip closure under flap with press studs
  • Ankles adjustable by touch and close fastening, zip closure under flap at leg
  • Elasticated waist
With additional protection and lining at shoulders, elbows and knees, stitched seam and high grade extra protective thermal lining.
Mais informação do produto