Print…
Safety splash shields, Nalgene®
Fornecedor: Thermo Fisher Scientific
6356-0001
113-2801EA 208 EUR
113-2801
Safety splash shields, Nalgene®
Protecções Protecções de Segurança
PC/acetate with magnetic base, transparent, thickness: 3 mm. Safety splash shield with flexible arm attaches easily to benchtops, shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris.
  • Durable, scratch-resistant 3 mm thick polycarbonate shield provides clear field of vision
  • 35 cm adjustable arm is easy to manoeuvre
  • Magnetic base positions the shield horizontally, vertically or on an overhead surface
  • UV protection from 200 to 360 nanometres
Certificações: Complies with OSHA; blood-borneMais informação do produto