Print…
Cleanroom gloves, polychloroprene, sterile, TouchNTuff® DermaShield® 73-701
Fornecedor: Ansell
73-701/6 73-701/6.5 73-701/7 73-701/7.5 73-701/8 73-701/8.5 73-701/9
112-3591EA 805 EUR
112-3591 112-3592 112-3593 112-3594 112-3595 112-3596 112-3597
Cleanroom gloves, polychloroprene, sterile, TouchNTuff® DermaShield® 73-701
Luvas
This technically advanced glove is made of polychloroprene. While it provides the comfort and flexibility of natural rubber gloves, its neoprene composition offers better chemical resistance for optimal product and worker protection.
  • Sterile, powder-free
  • Free of latex proteins and accelerators, it minimises both Type I and Type IV allergies associated with natural rubber
  • Textured fingertips for improved grip
  • Straight cuff
Ideal for use in aseptic drug manufacturing, biotechnologies, medical device manufacturing and for the handling of cytostatics.