Print…
Disposable earplugs, Contours®
Fornecedor: Moldex
7400 7403 7450 7453
111-1166EA 52.7 EUR
111-1166 111-1167 111-1169 111-1170
Disposable earplugs, Contours®
Tampões de ouvidos Tampões de ouvidos descartáveis
These extremely comfortable earplugs made of soft polyurethane foam are available in two different sizes.
  • Anatomical shape easily adapts to the ear canal: Less pressure and more comfort
  • Large base helps provide proper insertion, fit, and removal
  • Bright colour for improved visibility
SNR: 35 dB
Certificações: EN 352-2:2002
Fornecimento: Supplied in a box or in the hygienic dispenser system MoldexStation.
Embalagem: In pairs.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable Maior segurança para a saúde e ambiente