Print…
Semimáscaras respiratórias com filtração de partículas, descartável, FFP1/FFP2/FFP3, X-plore® 1300
Fornecedor: Dräger
3951215 3951217 3951211 3951214 3951216 3951213 3951218 3951212
111-1040EA 67.3 EUR
111-1040 111-1044 111-0759 111-0760 111-1043 111-0907 111-1045 111-1545
Semimáscaras respiratórias com filtração de partículas, descartável, FFP1/FFP2/FFP3, X-plore® 1300
Máscaras respiratórias Máscaras Descartáveis Máscaras respiratórias para partículas
The X-plore® 1300 series offers models in all three EN protection classes: FFP1, FFP2 and FFP3. Available with or without CoolMax™ exhalation valve for reduced exhalation resistance and to avoid a build-up of heat. Odour models for tackling unpleasant odours are equipped with an additional activated carbon layer.
  • Special high performance filter media are brought together to form the CoolSafe™ filter material, ensuring a low level of exhalation resistance for users while delivering a high level of filter performance
  • Flexible nose clip ensures the product is
Mais informação do produto